Assay Details
Target Gene Details
Entrez Gene ID: | 23514 |
Gene Name: | scaffolding protein involved in DNA repair |
Gene Aliases: |
KIAA0146 |
Location: |
Chr.8:47260878-47736001 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1028036 | Chr.8:46620127 - 47343541 on Build GRCh38 | Gain |
|
| dgv7211n100 | Chr.8:47250820 - 47334731 on Build GRCh38 | Gain |
|
| esv3617091 | Chr.8:47242260 - 47325695 on Build GRCh38 | Gain |
|
| esv3576568 | Chr.8:47250805 - 47303222 on Build GRCh38 | Gain |
|
| esv3617092 | Chr.8:47253772 - 47303222 on Build GRCh38 | Gain |
|
| nsv1022487 | Chr.8:45935900 - 47480134 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map