Assay Details
Target Gene Details
Entrez Gene ID: | 104326051 |
Gene Name: | MAFA antisense RNA 1 |
Gene Aliases: |
TCONS_00014882 |
Location: |
Chr.8:143417679-143419150 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAFA-AS1 | NR_126358.1 | |||
| BE503758.1 | ||||
| BF448373.1 | ||||
| BX114021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470247 | Chr.8:143283403 - 143615483 on Build GRCh38 | Loss |
|
| esv3619107 | Chr.8:143340435 - 143431612 on Build GRCh38 | Gain |
|
| nsv1076121 | Chr.8:143417929 - 143418730 on Build GRCh38 | Deletion |
|
| nsv950012 | Chr.8:143358831 - 143442430 on Build GRCh38 | Deletion |
|
| nsv1119192 | Chr.8:143418030 - 143418630 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72703742] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map