Assay Details
Target Gene Details
Entrez Gene ID: | 389692 |
Gene Name: | MAF bZIP transcription factor A |
Gene Aliases: |
RIPE3b1, hMafA |
Location: |
Chr.8:143428060-143430432 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAFA | NM_201589.3 | 1 | 1338 | NP_963883.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2668424 | Chr.8:143428175 - 143431522 on Build GRCh38 | Deletion |
|
| nsv470247 | Chr.8:143283403 - 143615483 on Build GRCh38 | Loss |
|
| nsv612768 | Chr.8:143426598 - 143440218 on Build GRCh38 | Loss |
|
| esv3619107 | Chr.8:143340435 - 143431612 on Build GRCh38 | Gain |
|
| nsv950012 | Chr.8:143358831 - 143442430 on Build GRCh38 | Deletion |
|
| nsv824765 | Chr.8:143428058 - 143432127 on Build GRCh38 | Gain |
|
| nsv612769 | Chr.8:143429115 - 143459926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map