Assay Details
Target Gene Details
Entrez Gene ID: | 4956 |
Gene Name: | outer dense fiber of sperm tails 1 |
Gene Aliases: |
CT133, HSPB10, ODF, ODF2, ODF27, ODFP, ODFPG, ODFPGA, ODFPGB, RT7, SODF |
Location: |
Chr.8:102551620-102561017 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ODF1 | NM_024410.3 | NP_077721.2 | ||
| AK315276.1 | ||||
| BC104456.1 | AAI04457.1 | |||
| BC104457.1 | AAI04458.1 | |||
| DB457574.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3618226 | Chr.8:102558699 - 102560292 on Build GRCh38 | Loss |
|
| esv3576598 | Chr.8:102200959 - 102574604 on Build GRCh38 | Gain |
|
| esv2762761 | Chr.8:102480128 - 102613016 on Build GRCh38 | Gain |
|
| esv3618224 | Chr.8:102532605 - 102572593 on Build GRCh38 | Gain |
|
| esv33118 | Chr.8:102346339 - 103055944 on Build GRCh38 | Loss |
|
| esv2669040 | Chr.8:102558704 - 102560293 on Build GRCh38 | Deletion |
|
| nsv1024876 | Chr.8:102500557 - 102564430 on Build GRCh38 | Gain |
|
| esv22177 | Chr.8:102558710 - 102560297 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73701168] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map