Assay Details
Target Gene Details
Entrez Gene ID: | 25861 |
Gene Name: | whirlin |
Gene Aliases: |
CIP98, DFNB31, PDZD7B, USH2D, WI |
Location: |
Chr.9:114402078-114505509 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WHRN | NM_001083885.2 | NP_001077354.2 | ||
| NM_001173425.1 | NP_001166896.1 | |||
| NM_015404.3 | NP_056219.3 | |||
| XM_011518484.1 | XP_011516786.1 | |||
| XM_011518485.1 | XP_011516787.1 | |||
| XM_011518486.1 | XP_011516788.1 | |||
| XM_011518487.1 | XP_011516789.1 | |||
| XM_011518488.1 | XP_011516790.1 | |||
| XM_011518495.1 | XP_011516797.1 | |||
| AB040959.1 | ||||
| AK022854.1 | BAB14275.1 | |||
| AL110228.1 | CAB53685.2 | |||
| BC136416.1 | ||||
| BC142614.1 | ||||
| BC142684.1 | ||||
| BX329269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv12859n54 | Chr.9:114193988 - 114696195 on Build GRCh38 | Gain |
|
| dgv7706n100 | Chr.9:114182745 - 114697870 on Build GRCh38 | Gain |
|
| nsv831693 | Chr.9:114354170 - 114519615 on Build GRCh38 | Gain |
|
| dgv33e196 | Chr.9:114030664 - 114604363 on Build GRCh38 | Duplication |
|
| nsv615233 | Chr.9:114167820 - 114486199 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map