Assay Details
Target Gene Details
Entrez Gene ID: | 259 |
Gene Name: | alpha-1-microglobulin/bikunin precursor |
Gene Aliases: |
A1M, EDC1, HCP, HI30, IATIL, ITI, ITIL, ITILC, UTI |
Location: |
Chr.9:114060127-114078472 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMBP | NM_001633.3 | NP_001624.1 | ||
| AK290837.1 | ||||
| AK303795.1 | ||||
| AK310776.1 | ||||
| AK310791.1 | ||||
| AY544123.1 | AAT11154.1 | |||
| BC041593.1 | AAH41593.1 | |||
| X04225.1 | CAA27803.1 | |||
| X04494.1 | CAA28182.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761285 | Chr.9:114049617 - 114340988 on Build GRCh38 | Gain |
|
| nsv528822 | Chr.9:114053497 - 114104774 on Build GRCh38 | Gain |
|
| dgv33e196 | Chr.9:114030664 - 114604363 on Build GRCh38 | Duplication |
|
| esv3621549 | Chr.9:113836116 - 114331078 on Build GRCh38 | Gain |
|
| dgv945n27 | Chr.9:114048610 - 114104774 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map