Assay Details
Target Gene Details
Entrez Gene ID: | 1029 |
Gene Name: | cyclin dependent kinase inhibitor 2A |
Gene Aliases: |
ARF, CDK4I, CDKN2, CMM2, INK4, INK4A, MLM, MTS-1, MTS1, P14, P14ARF, P16, P16-INK4A, P16INK4, P16INK4A, P19, P19ARF, TP16 |
Location: |
Chr.9:21967752-21995043 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN2A | NM_000077.4 | NP_000068.1 | ||
| NM_001195132.1 | NP_001182061.1 | |||
| NM_058195.3 | NP_478102.2 | |||
| NM_058197.4 | NP_478104.2 | |||
| XM_005251343.1 | XP_005251400.1 | |||
| XM_011517675.2 | 3 | 1020 | XP_011515977.1 | |
| XM_011517676.2 | XP_011515978.1 | |||
| XM_011517679.1 | XP_011515981.1 | |||
| AF115544.1 | AAD11437.1 | |||
| AI859822.1 | ||||
| AL582909.3 | ||||
| BC015960.2 | ||||
| BC021998.2 | ||||
| BI258230.1 | ||||
| BQ012762.1 | ||||
| DQ318021.1 | ||||
| JQ694043.1 | ||||
| JQ694044.1 | ||||
| JQ694045.1 | ||||
| L27211.1 | AAA92554.1 | |||
| S78535.1 | AAC60649.1 | |||
| U26727.1 | AAA82236.1 | |||
| U38945.1 | AAB01737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018514 | Chr.9:21944276 - 22075327 on Build GRCh38 | Gain |
|
| esv2764150 | Chr.9:20659506 - 22349184 on Build GRCh38 | Gain+Loss |
|
| nsv8426 | Chr.9:21763811 - 22176201 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72547291] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map