Assay Details
Target Gene Details
Entrez Gene ID: | 10783 |
Gene Name: | NIMA related kinase 6 |
Gene Aliases: |
SID6-1512 |
Location: |
Chr.9:124257606-124352442 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEK6 | NM_001145001.2 | NP_001138473.1 | ||
| NM_001166167.1 | NP_001159639.1 | |||
| NM_001166168.1 | NP_001159640.1 | |||
| NM_014397.5 | NP_055212.2 | |||
| XM_005251664.1 | XP_005251721.1 | |||
| AB026289.1 | BAA85045.1 | |||
| AF087909.1 | AAG13417.1 | |||
| AK313071.1 | ||||
| BC000101.2 | AAH00101.2 | |||
| BC004174.2 | AAH04174.2 | |||
| BC004209.2 | AAH04209.2 | |||
| BC012761.2 | AAH12761.1 | |||
| BM083872.1 | ||||
| BP265230.1 | ||||
| BP279761.1 | ||||
| CF994196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831719 | Chr.9:124093337 - 124279941 on Build GRCh38 | Gain |
|
| nsv519767 | Chr.9:124247983 - 124364893 on Build GRCh38 | Loss |
|
| nsv471322 | Chr.9:124247835 - 124357163 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map