Assay Details
Target Gene Details
Entrez Gene ID: | 10783 |
Gene Name: | NIMA related kinase 6 |
Gene Aliases: |
SID6-1512 |
Location: |
Chr.9:124257606-124352442 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEK6 | NM_001166167.1 | 1 | 89 | NP_001159639.1 |
| BC000101.2 | 1 | 88 | AAH00101.2 | |
| BM083872.1 | 1 | 89 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831719 | Chr.9:124093337 - 124279941 on Build GRCh38 | Gain |
|
| nsv831720 | Chr.9:124093669 - 124262769 on Build GRCh38 | Loss |
|
| nsv519767 | Chr.9:124247983 - 124364893 on Build GRCh38 | Loss |
|
| nsv6711 | Chr.9:124217250 - 124262051 on Build GRCh38 | Deletion |
|
| nsv825074 | Chr.9:124256071 - 124259412 on Build GRCh38 | Gain |
|
| nsv471322 | Chr.9:124247835 - 124357163 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map