Assay Details
Target Gene Details
Entrez Gene ID: | 51198 |
Gene Name: | CDKN2A antisense RNA 1 (head to head) |
Gene Aliases: |
C9orf53, bA149I2.3 |
Location: |
Chr.9:21967139-21967754 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN2A-AS1 | NR_024274.1 | 1 | 173 | |
| AF211119.1 | 1 | 191 | AAF22974.1 | |
| BX091351.1 | 1 | 173 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018514 | Chr.9:21944276 - 22075327 on Build GRCh38 | Gain |
|
| esv2764150 | Chr.9:20659506 - 22349184 on Build GRCh38 | Gain+Loss |
|
| nsv8426 | Chr.9:21763811 - 22176201 on Build GRCh38 | Loss |
|

Genomic Map