Assay Details
Target Gene Details
Entrez Gene ID: | 375748 |
Gene Name: | ERCC excision repair 6 like 2 |
Gene Aliases: |
BMFS2, C9orf102, RAD26L, SR278 |
Location: |
Chr.9:95875618-96041092 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERCC6L2 | XM_011518645.2 | XP_011516947.1 | ||
| AK308131.1 |
Target Gene Details
Entrez Gene ID: | 100188953 |
Gene Name: | long intergenic non-protein coding RNA 92 |
Gene Aliases: |
NCRNA00092, bA346B7.1 |
Location: |
Chr.9:96019732-96021755 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00092 | NR_024129.1 | 2 | 774 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524266 | Chr.9:96018324 - 96041992 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map