Assay Details
Target Gene Details
Entrez Gene ID: | 774 |
Gene Name: | calcium voltage-gated channel subunit alpha1 B |
Gene Aliases: |
BIII, CACNL1A5, CACNN, Cav2.2, DYT23 |
Location: |
Chr.9:137877789-138124624 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA1B | NM_000718.3 | NP_000709.1 | ||
| NM_001243812.1 | NP_001230741.1 | |||
| XM_011518991.2 | XP_011517293.1 | |||
| XM_011518992.2 | XP_011517294.1 | |||
| XM_011518993.2 | XP_011517295.1 | |||
| XM_011518994.2 | XP_011517296.1 | |||
| XM_017015115.1 | XP_016870604.1 | |||
| XM_017015116.1 | XP_016870605.1 | |||
| XM_017015117.1 | XP_016870606.1 | |||
| XM_017015118.1 | XP_016870607.1 | |||
| XM_017015119.1 | XP_016870608.1 | |||
| XM_017015120.1 | XP_016870609.1 | |||
| AB209467.1 | BAD92704.1 | |||
| M94172.1 | AAA51897.1 | |||
| M94173.1 | AAA51898.1 |
Target Gene Details
Entrez Gene ID: | 101928786 |
Gene Name: | uncharacterized LOC101928786 |
Gene Aliases: |
- |
Location: |
Chr.9:138040503-138045071 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928786 | NR_121582.1 | |||
| AK126935.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1043683 | Chr.9:137996262 - 138061095 on Build GRCh38 | Gain |
|
| esv2759722 | Chr.9:137991111 - 138262981 on Build GRCh38 | Gain |
|
| nsv469920 | Chr.9:137971737 - 138133487 on Build GRCh38 | Gain |
|
| nsv1041587 | Chr.9:137983555 - 138200932 on Build GRCh38 | Gain |
|
| nsv831764 | Chr.9:137989794 - 138171401 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map