Assay Details
Target Gene Details
Entrez Gene ID: | 100132159 |
Gene Name: | CALML3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:5514244-5526246 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALML3-AS1 | NR_120496.1 | |||
| NR_120497.1 | ||||
| AK128534.1 | BAC87486.1 | |||
| DA220455.1 | ||||
| DA770774.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831774 | Chr.10:5421593 - 5615458 on Build GRCh38 | Gain |
|
| nsv549886 | Chr.10:5163474 - 5590097 on Build GRCh38 | Loss |
|
| nsv523491 | Chr.10:5500146 - 5678104 on Build GRCh38 | Gain |
|
| nsv1051283 | Chr.10:5463511 - 5577978 on Build GRCh38 | Gain |
|
| esv2759729 | Chr.10:5515061 - 5762817 on Build GRCh38 | Gain |
|

Genomic Map