Assay Details
Target Gene Details
Entrez Gene ID: | 29974 |
Gene Name: | APOBEC1 complementation factor |
Gene Aliases: |
ACF, ACF64, ACF65, APOBEC1CF, ASP |
Location: |
Chr.10:50799409-50885681 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A1CF | NM_001198818.1 | 14 | 8288 | NP_001185747.1 |
| NM_001198819.1 | 15 | 8455 | NP_001185748.1 | |
| NM_001198820.1 | 14 | 8338 | NP_001185749.1 | |
| NM_014576.3 | 13 | 8195 | NP_055391.2 | |
| NM_138932.2 | 13 | 8219 | NP_620310.1 | |
| NM_138933.2 | 13 | 8290 | NP_620311.1 | |
| XM_005269718.2 | 14 | 8264 | XP_005269775.1 | |
| XM_005269720.3 | 12 | 8177 | XP_005269777.1 | |
| XM_011539729.2 | 12 | 8156 | XP_011538031.1 | |
| XM_011539730.2 | 14 | 8242 | XP_011538032.1 | |
| XM_017016160.1 | 12 | 8153 | XP_016871649.1 | |
| AK130022.1 | 1 | 1539 | ||
| CR749831.1 | 1 | 5486 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035197 | Chr.10:50686822 - 50862589 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map