Assay Details
Target Gene Details
Entrez Gene ID: | 79812 |
Gene Name: | multimerin 2 |
Gene Aliases: |
EMILIN-3, EMILIN3, ENDOGLYX-1 |
Location: |
Chr.10:86935540-86971311 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMRN2 | XM_006717970.3 | XP_006718033.1 |
Target Gene Details
Entrez Gene ID: | 6623 |
Gene Name: | synuclein gamma |
Gene Aliases: |
BCSG1, SR |
Location: |
Chr.10:86958531-86963260 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNCG | NM_003087.2 | NP_003078.2 | ||
| XM_011540105.2 | XP_011538407.1 | |||
| AF010126.1 | AAB64109.1 | |||
| AF017256.1 | AAC36550.1 | |||
| AF411524.1 | AAL05870.1 | |||
| AK223367.1 | BAD97087.1 | |||
| AK312141.1 | ||||
| BC014098.2 | AAH14098.1 | |||
| BP346786.1 | ||||
| CR541788.1 | CAG46587.1 | |||
| CR541790.1 | CAG46589.1 | |||
| EU019993.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428236 | Chr.10:86866859 - 87550005 on Build GRCh38 | Loss |
|
| esv33376 | Chr.10:86956341 - 86964954 on Build GRCh38 | Loss |
|
| nsv517487 | Chr.10:86899290 - 86973435 on Build GRCh38 | Gain+Loss |
|
| esv2759774 | Chr.10:86755301 - 87550005 on Build GRCh38 | Gain+Loss |
|
| nsv470969 | Chr.10:86936624 - 86972907 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map