Assay Details
Target Gene Details
Entrez Gene ID: | 8031 |
Gene Name: | nuclear receptor coactivator 4 |
Gene Aliases: |
ARA70, ELE1, PTC3, RFG |
Location: |
Chr.10:46005088-46030714 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCOA4 | NM_001145260.1 | NP_001138732.1 | ||
| NM_001145261.1 | NP_001138733.1 | |||
| NM_001145262.1 | 1 | 141 | NP_001138734.1 | |
| NM_001145263.1 | NP_001138735.1 | |||
| NM_005437.3 | NP_005428.1 | |||
| AK129911.1 | ||||
| AK292480.1 | 1 | 141 | ||
| AK293978.1 | ||||
| AK302794.1 | ||||
| AK304123.1 | ||||
| AK310849.1 | ||||
| AK312325.1 | ||||
| AL162047.1 | CAB82390.1 | |||
| BC001562.1 | AAH01562.1 | |||
| BC012736.2 | AAH12736.1 | |||
| DA415606.1 | ||||
| DA860108.1 | ||||
| DB075100.1 | ||||
| X77548.1 | CAA54673.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041519 | Chr.10:45991149 - 46080365 on Build GRCh38 | Gain |
|
| esv3623188 | Chr.10:46001576 - 46021276 on Build GRCh38 | Gain |
|
| esv3577172 | Chr.10:45959639 - 46146408 on Build GRCh38 | Gain |
|
| nsv508583 | Chr.10:45881821 - 46146976 on Build GRCh38 | Deletion |
|
| nsv1042912 | Chr.10:46009162 - 46116183 on Build GRCh38 | Loss |
|
| esv3623185 | Chr.10:45995694 - 46106028 on Build GRCh38 | Gain |
|
| nsv521269 | Chr.10:46001360 - 46080288 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map