Assay Details
Target Gene Details
Entrez Gene ID: | 389932 |
Gene Name: | aldo-keto reductase family 1 member C6, pseudogene |
Gene Aliases: |
TAKR |
Location: |
Chr.10:4871667-4916273 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR1C6P | NR_026743.1 | |||
| AB037902.1 | BAA92887.1 | |||
| AB037903.1 | BAA92888.1 | |||
| CB111913.1 |
Target Gene Details
Entrez Gene ID: | 83592 |
Gene Name: | aldo-keto reductase family 1 member E2 |
Gene Aliases: |
AKR1CL2, AKRDC1, LoopADR, hTSP, htAKR |
Location: |
Chr.10:4824554-4908421 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKR1E2 | XM_017016744.1 | XP_016872233.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549881 | Chr.10:4859606 - 5277716 on Build GRCh38 | Loss |
|
| nsv1045155 | Chr.10:4830047 - 5169355 on Build GRCh38 | Loss |
|
| nsv1042940 | Chr.10:4874422 - 4932389 on Build GRCh38 | Loss |
|
| nsv1050246 | Chr.10:4885236 - 5169355 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77652571] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map