Assay Details
Target Gene Details
Entrez Gene ID: | 3778 |
Gene Name: | potassium calcium-activated channel subfamily M alpha 1 |
Gene Aliases: |
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1 |
Location: |
Chr.10:76869601-77637819 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Target Gene Details
Entrez Gene ID: | 101929286 |
Gene Name: | KCNMA1 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.10:77350872-77376613 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNMA1-AS3 | NR_126365.1 | |||
| AA454124.1 | ||||
| AW510993.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759769 | Chr.10:77351633 - 77833713 on Build GRCh38 | Gain+Loss |
|
| nsv831919 | Chr.10:77292047 - 77470200 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map