Assay Details
Target Gene Details
Entrez Gene ID: | 340654 |
Gene Name: | lipase family member M |
Gene Aliases: |
LIPL3, bA304I5.1 |
Location: |
Chr.10:88802730-88820857 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPM | NM_001128215.1 | NP_001121687.1 | ||
| XM_011539748.2 | XP_011538050.1 | |||
| XM_011539751.2 | XP_011538053.1 | |||
| XM_011539752.2 | XP_011538054.1 | |||
| BC157888.1 | ||||
| BC171908.1 | ||||
| EF426484.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551840 | Chr.10:88718726 - 88841108 on Build GRCh38 | Gain |
|
| esv3624140 | Chr.10:88791335 - 88872446 on Build GRCh38 | Loss |
|
| nsv831942 | Chr.10:88804940 - 88970595 on Build GRCh38 | Gain |
|
| nsv1043015 | Chr.10:88707310 - 88833935 on Build GRCh38 | Gain |
|
| nsv831941 | Chr.10:88650731 - 88827575 on Build GRCh38 | Gain |
|
| esv3624139 | Chr.10:88724980 - 89003307 on Build GRCh38 | Gain |
|
| nsv7484 | Chr.10:88797991 - 88839915 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map