Assay Details
Target Gene Details
Entrez Gene ID: | 55760 |
Gene Name: | DEAH-box helicase 32 (putative) |
Gene Aliases: |
DDX32, DHLP1 |
Location: |
Chr.10:125836337-125896483 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX32 | NM_018180.2 | NP_060650.2 | ||
| XM_017016404.1 | XP_016871893.1 | |||
| XM_017016405.1 | XP_016871894.1 | |||
| AF427340.1 | AAL26550.1 | |||
| AK001556.1 | BAA91754.1 | |||
| AK001751.1 | BAA91882.1 | |||
| AK315430.1 | ||||
| AY064250.1 | AAL55441.1 | |||
| BC002473.2 | ||||
| BC068471.1 | AAH68471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759788 | Chr.10:125765331 - 126098133 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map