Assay Details
Target Gene Details
Entrez Gene ID: | 57178 |
Gene Name: | zinc finger MIZ-type containing 1 |
Gene Aliases: |
MIZ, RAI17, TRAFIP10, ZIMP10, hZIMP10 |
Location: |
Chr.10:79068994-79316528 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZMIZ1 | NM_020338.3 | NP_065071.1 | ||
| XM_005269988.3 | XP_005270045.1 | |||
| XM_006717923.3 | XP_006717986.1 | |||
| XM_006717924.3 | XP_006717987.1 | |||
| XM_006717925.3 | XP_006717988.1 | |||
| BC007288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv467384 | Chr.10:78349364 - 79336832 on Build GRCh38 | Gain |
|
| nsv551571 | Chr.10:79068826 - 79070719 on Build GRCh38 | Gain+Loss |
|
| nsv1131001 | Chr.10:79066743 - 79070943 on Build GRCh38 | Deletion |
|
| dgv1274n54 | Chr.10:79068707 - 79070719 on Build GRCh38 | Loss |
|
| esv26238 | Chr.10:79066222 - 79071664 on Build GRCh38 | Loss |
|
| nsv469807 | Chr.10:79043070 - 79212669 on Build GRCh38 | Loss |
|
| nsv1118179 | Chr.10:79070043 - 79071043 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map