Assay Details
Target Gene Details
Entrez Gene ID: | 119385 |
Gene Name: | ArfGAP with GTPase domain, ankyrin repeat and PH domain 11 |
Gene Aliases: |
- |
Location: |
Chr.10:86970741-87010203 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGAP11 | NM_133447.1 | NP_597704.1 | ||
| AK097165.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428236 | Chr.10:86866859 - 87550005 on Build GRCh38 | Loss |
|
| esv2759774 | Chr.10:86755301 - 87550005 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map