Assay Details
Target Gene Details
Entrez Gene ID: | 57705 |
Gene Name: | WDFY family member 4 |
Gene Aliases: |
C10orf64 |
Location: |
Chr.10:48684815-48982956 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDFY4 | XM_005270004.3 | XP_005270061.1 | ||
| XM_011539988.2 | XP_011538290.1 | |||
| XM_017016465.1 | XP_016871954.1 | |||
| BC034937.1 | AAH34937.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map