Assay Details
Target Gene Details
Entrez Gene ID: | 91452 |
Gene Name: | acyl-CoA binding domain containing 5 |
Gene Aliases: |
- |
Location: |
Chr.10:27182624-27248674 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3622710 | Chr.10:27206246 - 27213897 on Build GRCh38 | Gain |
|
| esv3622708 | Chr.10:27138085 - 27696800 on Build GRCh38 | Gain |
|
| esv2664616 | Chr.10:26927640 - 27310597 on Build GRCh38 | Deletion |
|
| esv2422253 | Chr.10:27142437 - 27286441 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map