Assay Details
Target Gene Details
Entrez Gene ID: | 100128811 |
Gene Name: | GPR158 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:25158072-25176276 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR158-AS1 | NR_027333.1 | 2 | 1165 | |
| AK094758.1 | 2 | 1166 |
Target Gene Details
Entrez Gene ID: | 101929025 |
Gene Name: | long intergenic non-protein coding RNA 1516 |
Gene Aliases: |
- |
Location: |
Chr.10:25113058-25161233 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01516 | NR_120649.1 | |||
| DA753967.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75115732] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map