Assay Details
Target Gene Details
Entrez Gene ID: | 390003 |
Gene Name: | guanylate cyclase 2G, pseudogene |
Gene Aliases: |
GUCY2G |
Location: |
Chr.10:112308178-112356595 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCY2GP | NR_028134.1 | |||
| DY655407.1 | ||||
| DY655429.1 | ||||
| EG327740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624580 | Chr.10:112327745 - 112333443 on Build GRCh38 | Loss |
|
| nsv471669 | Chr.10:112220414 - 112371065 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs567514451] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map