Assay Details
Target Gene Details
Entrez Gene ID: | 91452 |
Gene Name: | acyl-CoA binding domain containing 5 |
Gene Aliases: |
- |
Location: |
Chr.10:27182624-27248674 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACBD5 | XM_017016884.1 | XP_016872373.1 | ||
| XM_017016885.1 | XP_016872374.1 | |||
| XM_017016887.1 | XP_016872376.1 | |||
| XM_017016888.1 | XP_016872377.1 | |||
| XM_017016889.1 | XP_016872378.1 | |||
| XM_017016890.1 | XP_016872379.1 | |||
| XM_017016891.1 | XP_016872380.1 | |||
| XM_017016893.1 | XP_016872382.1 | |||
| XM_017016895.1 | XP_016872384.1 | |||
| XM_017016896.1 | XP_016872385.1 | |||
| XM_017016898.1 | XP_016872387.1 | |||
| XM_017016900.1 | XP_016872389.1 | |||
| XM_017016901.1 | XP_016872390.1 | |||
| XM_017016904.1 | XP_016872393.1 | |||
| XM_017016905.1 | XP_016872394.1 |
Target Gene Details
Entrez Gene ID: | 84930 |
Gene Name: | microtubule associated serine/threonine kinase like |
Gene Aliases: |
GREATWALL, GW, GWL, MAST-L, THC2 |
Location: |
Chr.10:27154468-27187953 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MASTL | NM_001172303.2 | NP_001165774.1 | ||
| NM_001172304.2 | NP_001165775.1 | |||
| NM_001320756.1 | NP_001307685.1 | |||
| NM_001320757.1 | NP_001307686.1 | |||
| NM_032844.4 | NP_116233.2 | |||
| NR_135469.1 | ||||
| XM_005252631.3 | XP_005252688.1 | |||
| XM_005252632.3 | XP_005252689.1 | |||
| XM_006717519.3 | XP_006717582.1 | |||
| XM_011519751.2 | XP_011518053.1 | |||
| XM_017016852.1 | XP_016872341.1 | |||
| XM_017016853.1 | XP_016872342.1 | |||
| XM_017016856.1 | XP_016872345.1 | |||
| AK027719.1 | BAB55321.1 | |||
| AK074804.1 | BAC11218.1 | |||
| AK123004.1 | ||||
| AK225319.1 | ||||
| BC009107.1 | AAH09107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3622709 | Chr.10:27165209 - 27189102 on Build GRCh38 | Gain |
|
| esv2664616 | Chr.10:26927640 - 27310597 on Build GRCh38 | Deletion |
|
| esv3622708 | Chr.10:27138085 - 27696800 on Build GRCh38 | Gain |
|
| esv2422253 | Chr.10:27142437 - 27286441 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map