Assay Details
Target Gene Details
Entrez Gene ID: | 3778 |
Gene Name: | potassium calcium-activated channel subfamily M alpha 1 |
Gene Aliases: |
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1 |
Location: |
Chr.10:76869601-77637819 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Gene Details
Entrez Gene ID: | 101929286 |
Gene Name: | KCNMA1 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.10:77350872-77376613 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNMA1-AS3 | NR_126365.1 | 1 | 372 | |
| DB047348.1 | 1 | 372 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551551 | Chr.10:77261063 - 77359691 on Build GRCh38 | Loss |
|
| nsv831919 | Chr.10:77292047 - 77470200 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map