Assay Details
Target Gene Details
Entrez Gene ID: | 54777 |
Gene Name: | cilia and flagella associated protein 46 |
Gene Aliases: |
C10orf123, C10orf124, C10orf92, C10orf93, TTC40, bA288G11.4, bA288G11.5, bB137A17.2, bB137A17.3 |
Location: |
Chr.10:132808392-132942585 on Build GRCh38 |
Assay Gene Location: | Within Intron 44 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP46 | NM_001200049.2 | NP_001186978.2 | ||
| AL122111.1 | CAB59272.3 | |||
| BC034223.1 | AAH34223.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2750852 | Chr.10:132797506 - 132852920 on Build GRCh38 | Gain |
|
| nsv1042708 | Chr.10:132791887 - 132988338 on Build GRCh38 | Gain |
|
| nsv832033 | Chr.10:132743736 - 132870955 on Build GRCh38 | Loss |
|
| dgv995n100 | Chr.10:132825790 - 133019278 on Build GRCh38 | Gain |
|
| nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain |
|
| nsv552526 | Chr.10:132699017 - 133103793 on Build GRCh38 | Gain |
|
| nsv832034 | Chr.10:132761541 - 132940326 on Build GRCh38 | Loss |
|
| nsv832032 | Chr.10:132648722 - 132851966 on Build GRCh38 | Loss |
|
| nsv952700 | Chr.10:132702797 - 132851596 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114471525] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map