Assay Details
Target Gene Details
Entrez Gene ID: | 100505601 |
Gene Name: | PARD3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:34815768-34816422 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARD3-AS1 | NR_108043.1 | 2 | 181 | |
| BF511037.1 | 1 | 172 | ||
| BX089194.1 | 2 | 181 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv466860 | Chr.10:34383535 - 35056041 on Build GRCh38 | Gain |
|
| dgv76n27 | Chr.10:34731710 - 34865418 on Build GRCh38 | Gain |
|
| nsv521667 | Chr.10:34696031 - 34966937 on Build GRCh38 | Gain |
|
| dgv702n100 | Chr.10:34697315 - 34925688 on Build GRCh38 | Gain |
|
| dgv704n100 | Chr.10:34771494 - 34859811 on Build GRCh38 | Gain |
|
| esv3576338 | Chr.10:34734850 - 35002577 on Build GRCh38 | Gain |
|
| nsv24473 | Chr.10:34808014 - 34817122 on Build GRCh38 | Deletion |
|
| nsv550397 | Chr.10:34786889 - 34974998 on Build GRCh38 | Gain |
|
| nsv550392 | Chr.10:34695956 - 34985822 on Build GRCh38 | Gain |
|
| dgv703n100 | Chr.10:34729183 - 35050396 on Build GRCh38 | Gain |
|
| esv3622879 | Chr.10:34733295 - 34857006 on Build GRCh38 | Gain |
|

Genomic Map