Assay Details
Target Gene Details
Entrez Gene ID: | 9712 |
Gene Name: | USP6 N-terminal like |
Gene Aliases: |
RNTRE, TRE2NL, USP6NL-IT1 |
Location: |
Chr.10:11460510-11611789 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP6NL | NM_001080491.3 | NP_001073960.1 | ||
| NM_014688.3 | NP_055503.1 | |||
| XM_006717542.3 | XP_006717605.1 | |||
| XM_011519762.2 | XP_011518064.1 | |||
| XM_011519763.2 | XP_011518065.1 | |||
| XM_017016971.1 | XP_016872460.1 | |||
| XM_017016972.1 | XP_016872461.1 | |||
| XM_017016973.1 | XP_016872462.1 | |||
| AB449916.1 | ||||
| AK292944.1 | ||||
| BC042943.2 | AAH42943.1 | |||
| D13644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3578632 | Chr.10:11471721 - 11478978 on Build GRCh38 | Loss |
|
| esv2658637 | Chr.10:11478400 - 11480469 on Build GRCh38 | Deletion |
|
| esv3622339 | Chr.10:11478380 - 11480469 on Build GRCh38 | Loss |
|
| nsv831784 | Chr.10:11435937 - 11614982 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map