Assay Details
Target Gene Details
Entrez Gene ID: | 29118 |
Gene Name: | DEAD-box helicase 25 |
Gene Aliases: |
GRTH |
Location: |
Chr.11:125903275-125923110 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX25 | NM_013264.4 | NP_037396.3 | ||
| XM_011542790.2 | XP_011541092.1 | |||
| XM_011542791.2 | XP_011541093.1 | |||
| XM_017017626.1 | XP_016873115.1 | |||
| AF155140.2 | AAF21371.2 | |||
| AK295121.1 | ||||
| AK312772.1 | ||||
| BC035388.1 | ||||
| BC050360.1 | ||||
| CD359894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv248n67 | Chr.11:125843561 - 125919531 on Build GRCh38 | Gain |
|
| nsv556525 | Chr.11:125690285 - 125971104 on Build GRCh38 | Gain |
|
| esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map