Assay Details
Target Gene Details
Entrez Gene ID: | 29125 |
Gene Name: | chromosome 11 open reading frame 21 |
Gene Aliases: |
- |
Location: |
Chr.11:2296277-2301913 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C11orf21 | NM_001142946.1 | NP_001136418.1 | ||
| NR_024621.1 | ||||
| XM_011520034.1 | XP_011518336.1 | |||
| AB029488.1 | BAA89482.1 | |||
| AK310172.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469928 | Chr.11:2294174 - 2473462 on Build GRCh38 | Loss |
|
| nsv553082 | Chr.11:2296721 - 2300906 on Build GRCh38 | Loss |
|
| nsv553081 | Chr.11:2291007 - 2312445 on Build GRCh38 | Loss |
|
| nsv553080 | Chr.11:2289922 - 2301432 on Build GRCh38 | Loss |
|
| nsv467645 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain |
|
| nsv951285 | Chr.11:2256071 - 2310670 on Build GRCh38 | Deletion |
|

Genomic Map