Assay Details
Target Gene Details
Entrez Gene ID: | 10077 |
Gene Name: | tetraspanin 32 |
Gene Aliases: |
ART1, PHEMX, PHMX, TSSC6 |
Location: |
Chr.11:2299480-2318201 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469928 | Chr.11:2294174 - 2473462 on Build GRCh38 | Loss |
|
| esv3625125 | Chr.11:2303216 - 2328448 on Build GRCh38 | Gain |
|
| nsv467645 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map