Assay Details
Target Gene Details
Entrez Gene ID: | 6833 |
Gene Name: | ATP binding cassette subfamily C member 8 |
Gene Aliases: |
ABC36, HHF1, HI, HRINS, MRP8, PHHI, SUR, SUR1, SUR1delta2, TNDM2 |
Location: |
Chr.11:17392885-17476870 on Build GRCh38 |
Assay Gene Location: | Within Intron 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCC8 | NM_000352.4 | NP_000343.2 | ||
| NM_001287174.1 | NP_001274103.1 | |||
| XM_011520331.2 | XP_011518633.1 | |||
| XM_017018197.1 | XP_016873686.1 | |||
| XM_017018198.1 | XP_016873687.1 | |||
| XM_017018199.1 | XP_016873688.1 | |||
| XM_017018200.1 | XP_016873689.1 | |||
| XM_017018201.1 | XP_016873690.1 | |||
| XM_017018202.1 | XP_016873691.1 | |||
| XM_017018204.1 | XP_016873693.1 | |||
| AB209084.1 | BAD92321.1 | |||
| AF087138.1 | AAC36724.1 | |||
| BM352492.1 | ||||
| L40625.1 | AAA99227.1 | |||
| L78207.1 | AAB02278.1 | |||
| U63421.1 | AAB36699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553567 | Chr.11:16890704 - 17946708 on Build GRCh38 | Gain |
|
| dgv59n21 | Chr.11:17379587 - 17413667 on Build GRCh38 | Loss |
|
| nsv553584 | Chr.11:17386478 - 17418926 on Build GRCh38 | Loss |
|
| nsv469895 | Chr.11:17346723 - 17562673 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map