Assay Details
Target Gene Details
Entrez Gene ID: | 57558 |
Gene Name: | ubiquitin specific peptidase 35 |
Gene Aliases: |
- |
Location: |
Chr.11:78188812-78233651 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP35 | NM_020798.2 | 6 | 1501 | NP_065849.1 |
| XM_011545187.2 | 6 | 644 | XP_011543489.1 | |
| XM_011545188.2 | 4 | 427 | XP_011543490.1 | |
| XM_017018052.1 | 6 | 1498 | XP_016873541.1 | |
| XM_017018053.1 | 5 | 493 | XP_016873542.1 | |
| AB037793.1 | 3 | 964 | BAA92610.1 | |
| AJ586137.1 | 5 | 1142 | CAE51937.1 | |
| AK056231.1 | ||||
| AK301048.1 | ||||
| AK307875.1 | 5 | 727 | ||
| BC131489.1 | 4 | 464 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain |
|
| nsv516503 | Chr.11:78145131 - 78209884 on Build GRCh38 | Loss |
|
| nsv1132346 | Chr.11:78199654 - 78201354 on Build GRCh38 | Duplication |
|
| nsv527066 | Chr.11:78172809 - 78252087 on Build GRCh38 | Gain |
|
| nsv1036793 | Chr.11:78198402 - 78265821 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map