Assay Details
Target Gene Details
Entrez Gene ID: | 26011 |
Gene Name: | teneurin transmembrane protein 4 |
Gene Aliases: |
Doc4, ETM5, ODZ4, TNM4, Ten-M4 |
Location: |
Chr.11:78653283-79441019 on Build GRCh38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TENM4 | NM_001098816.2 | 34 | 11806 | NP_001092286.2 |
| XM_011544933.2 | 17 | 8901 | XP_011543235.1 | |
| XM_017017525.1 | 33 | 12077 | XP_016873014.1 | |
| XM_017017526.1 | 31 | 12982 | XP_016873015.1 | |
| AB037723.3 | 19 | 9084 | BAA92540.3 | |
| AK023935.1 | 1 | 493 | ||
| AL080120.1 | 1 | 3589 | CAB45719.1 | |
| BX640737.1 | 8 | 7233 | CAE45850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483084 | Chr.11:78645595 - 78818126 on Build GRCh38 | Loss |
|
| nsv468643 | Chr.11:78629643 - 78671072 on Build GRCh38 | Gain |
|
| dgv211e214 | Chr.11:78524451 - 78661281 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map