Assay Details
Target Gene Details
Entrez Gene ID: | 57571 |
Gene Name: | carnosine synthase 1 |
Gene Aliases: |
ATPGD1 |
Location: |
Chr.11:67415678-67425607 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARNS1 | NM_001166222.1 | NP_001159694.1 | ||
| NM_020811.1 | NP_065862.1 | |||
| XM_011545189.2 | XP_011543491.1 | |||
| XM_011545191.2 | 1 | 600 | XP_011543493.1 | |
| XM_017018054.1 | 2 | 302 | XP_016873543.1 | |
| AB037815.1 | 1 | 196 | ||
| AK294035.1 | ||||
| AK295331.1 | 2 | 302 | ||
| EF560746.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516453 | Chr.11:67397024 - 67418529 on Build GRCh38 | Loss |
|
| dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion |
|
| nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss |
|
| nsv555255 | Chr.11:67410326 - 67423978 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map