Assay Details
Target Gene Details
Entrez Gene ID: | 10077 |
Gene Name: | tetraspanin 32 |
Gene Aliases: |
ART1, PHEMX, PHMX, TSSC6 |
Location: |
Chr.11:2299480-2318201 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN32 | NM_139022.2 | NP_620591.3 | ||
| XM_005252719.4 | 9 | 1112 | XP_005252776.1 | |
| XM_011519815.2 | 8 | 1034 | XP_011518117.2 | |
| XM_011519818.2 | XP_011518120.2 | |||
| XM_011519819.1 | 9 | 1000 | XP_011518121.1 | |
| XM_011519822.1 | XP_011518124.1 | |||
| XM_017017064.1 | 8 | 1584 | XP_016872553.1 | |
| XM_017017065.1 | XP_016872554.1 | |||
| XM_017017066.1 | 7 | 1500 | XP_016872555.1 | |
| XM_017017067.1 | 9 | 1004 | XP_016872556.1 | |
| XM_017017068.1 | XP_016872557.1 | |||
| AF125569.1 | AAD23580.1 | |||
| AF176070.1 | AAG15840.1 | |||
| AF176071.1 | AAG15841.1 | |||
| AK128812.1 | ||||
| AY039001.1 | AAK84431.1 | |||
| AY303780.1 | AAP76185.1 | |||
| BC016693.1 | AAH16693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469928 | Chr.11:2294174 - 2473462 on Build GRCh38 | Loss |
|
| esv3625125 | Chr.11:2303216 - 2328448 on Build GRCh38 | Gain |
|
| nsv467645 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map