Assay Details
Target Gene Details
Entrez Gene ID: | 283219 |
Gene Name: | potassium channel tetramerization domain containing 21 |
Gene Aliases: |
KCASH2 |
Location: |
Chr.11:78171243-78188854 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD21 | XM_005273925.3 | 1 | 90 | XP_005273982.1 |
Target Gene Details
Entrez Gene ID: | 57558 |
Gene Name: | ubiquitin specific peptidase 35 |
Gene Aliases: |
- |
Location: |
Chr.11:78188812-78233651 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP35 | NM_020798.2 | NP_065849.1 | ||
| XM_017018052.1 | XP_016873541.1 | |||
| AK056231.1 | ||||
| AK301048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain |
|
| nsv516503 | Chr.11:78145131 - 78209884 on Build GRCh38 | Loss |
|
| nsv527066 | Chr.11:78172809 - 78252087 on Build GRCh38 | Gain |
|
| esv33345 | Chr.11:78177662 - 78190142 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map