Assay Details
Target Gene Details
Entrez Gene ID: | 338667 |
Gene Name: | V-set and immunoglobulin domain-containing protein 10-like |
Gene Aliases: |
- |
Location: |
Chr.11:125949191-125956319 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC338667 | XM_017018652.1 | XP_016874141.1 | ||
| XM_017018653.1 | XP_016874142.1 | |||
| XM_017018654.1 | XP_016874143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv535 | Chr.11:125918491 - 125963371 on Build GRCh38 | Deletion |
|
| nsv556525 | Chr.11:125690285 - 125971104 on Build GRCh38 | Gain |
|
| nsv516919 | Chr.11:125942455 - 125955813 on Build GRCh38 | Loss |
|
| esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss |
|

Genomic Map