Assay Details
Target Gene Details
Entrez Gene ID: | 9024 |
Gene Name: | BR serine/threonine kinase 2 |
Gene Aliases: |
C11orf7, PEN11B, SAD1, SADA, STK29 |
Location: |
Chr.11:1389899-1462689 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951279 | Chr.11:1337871 - 1491070 on Build GRCh38 | Deletion |
|
| nsv552984 | Chr.11:1412088 - 1417774 on Build GRCh38 | Gain |
|
| nsv552985 | Chr.11:1412139 - 1417671 on Build GRCh38 | Loss |
|
| esv2669189 | Chr.11:864834 - 1489637 on Build GRCh38 | Deletion |
|
| dgv10n68 | Chr.11:1363751 - 1500210 on Build GRCh38 | Loss |
|
| nsv467642 | Chr.11:1383608 - 1428143 on Build GRCh38 | Loss |
|
| nsv552983 | Chr.11:1398714 - 1416873 on Build GRCh38 | Gain |
|
| esv2743878 | Chr.11:864732 - 1489652 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs182399375] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map