Assay Details
Target Gene Details
Entrez Gene ID: | 220074 |
Gene Name: | leucine rich transmembrane and O-methyltransferase domain containing |
Gene Aliases: |
CFAP111, DFNB63, LRRC51 |
Location: |
Chr.11:72079982-72110782 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRTOMT | NM_001145308.4 | 5 | 1107 | NP_001138780.1 |
| NM_001145309.3 | 7 | 1328 | NP_001138781.1 | |
| NM_001145310.3 | NP_001138782.1 | |||
| XM_017017356.1 | 2 | 1722 | XP_016872845.1 | |
| XM_017017357.1 | 1 | 2645 | XP_016872846.1 | |
| XM_017017358.1 | 1 | 2645 | XP_016872847.1 | |
| AF289588.1 | AAL55772.1 | |||
| AK302772.1 | 5 | 601 | ||
| EU627069.1 | 7 | 800 | ||
| EU627070.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv384 | Chr.11:72074236 - 72106174 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map