Assay Details
Target Gene Details
Entrez Gene ID: | 5002 |
Gene Name: | solute carrier family 22 member 18 |
Gene Aliases: |
BWR1A, BWSCR1A, HET, IMPT1, ITM, ORCTL2, SLC22A1L, TSSC5, p45-BWR1A |
Location: |
Chr.11:2899721-2925246 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC22A18 | NM_183233.2 | NP_899056.2 | ||
| BC015571.2 | AAH15571.1 |
Target Gene Details
Entrez Gene ID: | 5003 |
Gene Name: | solute carrier family 22 member 18 antisense |
Gene Aliases: |
BWR1B, BWSCR1B, ORCTL2S, SLC22A1LS, p27-BWR1B |
Location: |
Chr.11:2887344-2905504 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC22A18AS | NM_001302862.1 | NP_001289791.1 | ||
| NM_007105.3 | NP_009036.2 | |||
| XM_017017832.1 | XP_016873321.1 | |||
| XM_017017833.1 | XP_016873322.1 | |||
| XM_017017834.1 | XP_016873323.1 | |||
| AF037066.1 | AAC04789.1 | |||
| BC030237.1 | AAH30237.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759795 | Chr.11:2767814 - 2942382 on Build GRCh38 | Gain |
|
| nsv951292 | Chr.11:2792271 - 2927870 on Build GRCh38 | Deletion |
|
| nsv469930 | Chr.11:2831299 - 2924715 on Build GRCh38 | Loss |
|
| nsv832050 | Chr.11:2868540 - 2963987 on Build GRCh38 | Loss |
|
| nsv553111 | Chr.11:2842057 - 2904391 on Build GRCh38 | Loss |
|
| nsv553119 | Chr.11:2879188 - 2916401 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map