Assay Details
Target Gene Details
Entrez Gene ID: | 399909 |
Gene Name: | pecanex homolog 3 (Drosophila) |
Gene Aliases: |
PCNXL3 |
Location: |
Chr.11:65614311-65637439 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCNX3 | NM_032223.3 | NP_115599.2 | ||
| XM_005273987.4 | 11 | 4231 | XP_005274044.1 | |
| XM_005273990.4 | 11 | 4234 | XP_005274047.1 | |
| XM_006718549.3 | 11 | 4216 | XP_006718612.1 | |
| XM_006718551.3 | 11 | 4234 | XP_006718614.1 | |
| XM_011545024.2 | 12 | 3407 | XP_011543326.1 | |
| XM_011545025.2 | 11 | 4234 | XP_011543327.1 | |
| XM_011545026.2 | 5 | 1074 | XP_011543328.1 | |
| XM_011545027.1 | XP_011543329.1 | |||
| XM_011545028.2 | 11 | 4234 | XP_011543330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1969n54 | Chr.11:65568234 - 65663889 on Build GRCh38 | Loss |
|
| esv2761680 | Chr.11:65576857 - 65623846 on Build GRCh38 | Gain |
|
| nsv951018 | Chr.11:65525430 - 65662029 on Build GRCh38 | Deletion |
|

Genomic Map