Assay Details
Target Gene Details
Entrez Gene ID: | 266743 |
Gene Name: | neuronal PAS domain protein 4 |
Gene Aliases: |
Le-PAS, NXF, PASD10, bHLHe79 |
Location: |
Chr.11:66409158-66426707 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPAS4 | XM_017017537.1 | 2 | 627 | XP_016873026.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832193 | Chr.11:66352868 - 66511334 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map