Assay Details
Target Gene Details
Entrez Gene ID: | 120400 |
Gene Name: | neurexophilin and PC-esterase domain family member 1 |
Gene Aliases: |
FAM55A |
Location: |
Chr.11:114518946-114560244 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NXPE1 | XM_011542594.2 | 8 | 3263 | XP_011540896.1 |
XM_011542595.2 | 9 | 3375 | XP_011540897.1 | |
XM_011542596.2 | 9 | 3616 | XP_011540898.1 | |
XM_011542597.2 | 8 | 3504 | XP_011540899.1 | |
XM_011542598.2 | 7 | 3204 | XP_011540900.1 | |
XM_011542599.2 | 7 | 3445 | XP_011540901.1 | |
AK127883.1 | 1 | 113 |
Target Gene Details
Entrez Gene ID: | 120406 |
Gene Name: | neurexophilin and PC-esterase domain family member 2 |
Gene Aliases: |
FAM55B |
Location: |
Chr.11:114464276-114813702 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NXPE2 | XM_017017207.1 | XP_016872696.1 | ||
XM_017017209.1 | XP_016872698.1 | |||
XM_017017210.1 | XP_016872699.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759865 | Chr.11:114501563 - 114648403 on Build GRCh38 | Gain |
![]() ![]() ![]() |
