Assay Details
Target Gene Details
Entrez Gene ID: | 283358 |
Gene Name: | beta-1,4-N-acetyl-galactosaminyltransferase 3 |
Gene Aliases: |
- |
Location: |
Chr.12:460377-561892 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALNT3 | NM_173593.3 | NP_775864.3 | ||
| AB089940.1 | BAD02449.1 | |||
| CN400443.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628240 | Chr.12:468207 - 485357 on Build GRCh38 | Loss |
|
| esv2759875 | Chr.12:225844 - 881378 on Build GRCh38 | Gain+Loss |
|
| nsv1050781 | Chr.12:212187 - 674733 on Build GRCh38 | Loss |
|
| esv3628242 | Chr.12:470754 - 476578 on Build GRCh38 | Loss |
|
| esv3892121 | Chr.12:386051 - 528197 on Build GRCh38 | Gain |
|
| esv3628236 | Chr.12:417352 - 561839 on Build GRCh38 | Gain |
|
| nsv1040579 | Chr.12:466467 - 652394 on Build GRCh38 | Gain |
|
| nsv1046514 | Chr.12:444925 - 524251 on Build GRCh38 | Gain |
|
| dgv1332n100 | Chr.12:461781 - 508815 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111578988] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map