Assay Details
Target Gene Details
Entrez Gene ID: | 57666 |
Gene Name: | fibrosin like 1 |
Gene Aliases: |
- |
Location: |
Chr.12:132490440-132585188 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBRSL1 | NM_001142641.1 | NP_001136113.1 | ||
| XM_005266171.3 | XP_005266228.1 | |||
| XM_005266173.3 | XP_005266230.1 | |||
| XM_005266175.3 | XP_005266232.1 | |||
| XM_005266176.3 | XP_005266233.1 | |||
| XM_005266177.3 | XP_005266234.1 | |||
| XM_011534803.2 | XP_011533105.1 | |||
| XM_011534804.2 | XP_011533106.1 | |||
| XM_011534805.2 | XP_011533107.1 | |||
| XM_011534806.2 | XP_011533108.1 | |||
| XM_011534807.2 | XP_011533109.1 | |||
| XM_011534808.2 | XP_011533110.1 | |||
| XM_011534809.2 | XP_011533111.1 | |||
| XM_011534810.2 | XP_011533112.1 | |||
| XM_011534812.2 | XP_011533114.1 | |||
| XM_011534816.2 | XP_011533118.1 | |||
| AB046765.2 | BAB13371.2 |
Target Gene Details
Entrez Gene ID: | 100996573 |
Gene Name: | WAS/WASL-interacting protein family member 3-like |
Gene Aliases: |
- |
Location: |
Chr.12:132550729-132559465 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100996573 | XM_003846334.4 | 1 | 7891 | XP_003846382.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv455806 | Chr.12:132516994 - 132614597 on Build GRCh38 | Loss |
|
| nsv952512 | Chr.12:132192256 - 132711214 on Build GRCh38 | Deletion |
|
| nsv561105 | Chr.12:132540903 - 132584760 on Build GRCh38 | Loss |
|
| dgv3046n54 | Chr.12:132481702 - 132614597 on Build GRCh38 | Loss |
|
| nsv826569 | Chr.12:132553843 - 132588432 on Build GRCh38 | Gain |
|
| nsv826563 | Chr.12:132125383 - 132622340 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map