Assay Details
Target Gene Details
Entrez Gene ID: | 101927735 |
Gene Name: | uncharacterized LOC101927735 |
Gene Aliases: |
- |
Location: |
Chr.12:129208601-129212662 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927735 | NR_110058.1 |
Target Gene Details
Entrez Gene ID: | 121256 |
Gene Name: | transmembrane protein 132D |
Gene Aliases: |
MOLT, PPP1R153 |
Location: |
Chr.12:129071726-129903667 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM132D | NM_133448.2 | NP_597705.2 | ||
| AB061814.1 | BAC97794.1 | |||
| BC114626.1 | ||||
| BC114936.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832550 | Chr.12:129099732 - 129291248 on Build GRCh38 | Loss |
|
| nsv1040831 | Chr.12:128749443 - 129516029 on Build GRCh38 | Gain |
|
| esv3631162 | Chr.12:129183201 - 129249654 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map